Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Unr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144440.2 | NP_001137912.1 | 7 - 8 | 6460 | 64 | |
NM_001259740.2 | NP_001246669.1 | 9 - 10 | 6505 | 64 | ||
NM_001274650.1 | NP_001261579.1 | 7 - 8 | 3443 | 64 | ||
NM_001300054.1 | NP_001286983.1 | 7 - 8 | 6460 | 64 | ||
NM_139969.4 | NP_648226.2 | 9 - 10 | 6466 | 64 | ||
GenBank mRNA | BT133161.1 | - | 7 - 8 | 3439 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |