Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273836.1 | NP_001260765.1 | 1 - 2 | 784 | 82 | |
NM_057744.3 | NP_477092.1 | 1 - 2 | 784 | 82 | ||
GenBank mRNA | BT044167.1 | - | 1 - 2 | 763 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |