Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp4e2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299267.1 | NP_001286196.1 | 6 - 7 | 1325 | 88 | |
NM_057769.4 | NP_477117.2 | 5 - 6 | 1228 | 88 | ||
GenBank mRNA | AY058518.1 | - | 5 - 6 | 1204 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |