Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: onecut | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272136.1 | NP_001259065.1 | 2 - 3 | 1962 | 62 | |
NM_080103.3 | NP_524842.2 | 1 - 2 | 2654 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |