Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: or | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080532.4 | NP_536793.1 | 1 - 2 | 152 | 99 | |
GenBank mRNA | AF228056.1 | - | 1 - 2 | 102 | 99 | |
AY071026.1 | - | 1 - 2 | 146 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |