Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fok | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201926.2 | NP_001188855.1 | 1 - 2 | 488 | 59 | |
NM_080713.6 | NP_542444.1 | 1 - 2 | 488 | 59 | ||
GenBank mRNA | AY113608.1 | - | 1 - 2 | 486 | 59 | |
KX532092.1 | - | 1 - 2 | 152 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |