Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgant4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144307.2 | NP_001137779.1 | 1 - 2 | 570 | 101 | |
NM_164539.2 | NP_722910.2 | 1 - 2 | 702 | 101 | ||
GenBank mRNA | AF324752.1 | - | 1 - 2 | 502 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |