Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272373.1 | NP_001259302.1 | 4 - 5 | 660 | 63 | |
NM_176700.3 | NP_788873.1 | 4 - 5 | 660 | 63 | ||
GenBank mRNA | AF110233.1 | - | 4 - 5 | 646 | 63 | |
AJ009657.1 | - | 4 - 5 | 635 | 63 | ||
AY069388.1 | - | 4 - 5 | 636 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |