Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258777.1 | NP_001245706.1 | 1 - 2 | 2946 | 99 | |
NM_001258778.2 | NP_001245707.1 | 4 - 5 | 761 | 99 | ||
NM_001298390.1 | NP_001285319.1 | 2 - 3 | 2894 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |