Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TkR86C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104271.1 | NP_001097741.1 | 1 - 2 | 1130 | 72 | |
NM_079580.3 | NP_524304.2 | 1 - 2 | 719 | 72 | ||
GenBank mRNA | BT029262.1 | - | 1 - 2 | 1130 | 72 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Drosophila melanogaster |