Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170148.2 | NP_001163619.1 | 6 - 7 | 651 | 64 | |
NM_001260191.1 | NP_001247120.1 | 4 - 5 | 901 | 64 | ||
NM_001382175.1 | NP_001369014.1 | 6 - 7 | 938 | 64 | ||
NM_079636.4 | NP_524360.2 | 4 - 5 | 903 | 64 | ||
NM_206494.3 | NP_996216.1 | 6 - 7 | 651 | 64 | ||
GenBank mRNA | AY060414.1 | - | 4 - 5 | 883 | 64 | |
BT011469.1 | - | 6 - 7 | 649 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |