Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slmb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275872.1 | NP_001262801.1 | 4 - 5 | 1224 | 61 | |
NM_001275873.1 | NP_001262802.1 | 4 - 5 | 1224 | 61 | ||
NM_079706.3 | NP_524430.1 | 4 - 5 | 1224 | 61 | ||
GenBank mRNA | AF222923.1 | - | 4 - 5 | 1221 | 61 | |
AF222924.1 | - | 4 - 5 | 1103 | 61 | ||
AY118898.1 | - | 4 - 5 | 1192 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |