Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trbd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141674.3 | NP_649931.1 | 3 - 4 | 1303 | 62 | |
GenBank mRNA | AY058438.1 | - | 3 - 4 | 1265 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |