Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mtTFB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141714.4 | NP_649971.1 | 1 - 2 | 565 | 68 | |
GenBank mRNA | AY060616.1 | - | 1 - 2 | 541 | 68 | |
AY508412.1 | - | 1 - 2 | 541 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |