Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275748.1 | NP_001262677.1 | 2 - 3 | 1297 | 85 | |
NM_142387.3 | NP_650644.1 | 2 - 3 | 1297 | 85 | ||
GenBank mRNA | BT010086.1 | - | 2 - 3 | 1268 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |