Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EloA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_142878.4 | NP_651135.1 | 6 - 7 | 1708 | 63 | |
NM_170061.3 | NP_732846.1 | 6 - 7 | 1880 | 63 | ||
GenBank mRNA | AY061363.1 | - | 6 - 7 | 1684 | 63 | |
BT133197.1 | - | 6 - 7 | 1899 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |