Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pi4KIIalpha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001382052.1 | NP_001368956.1 | 4 - 5 | 994 | 60 | |
NM_001382053.1 | NP_001368992.1 | 3 - 4 | 320 | 60 | ||
NM_169063.4 | NP_730944.2 | 5 - 6 | 1398 | 60 | ||
NM_169064.2 | NP_730945.1 | 3 - 4 | 841 | 60 | ||
NM_206432.3 | NP_996154.1 | 5 - 6 | 1371 | 60 | ||
GenBank mRNA | AY102687.1 | - | 5 - 6 | 1344 | 60 | |
BT001304.1 | - | 3 - 4 | 841 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |