Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275559.1 | NP_001262488.1 | 2 | 580 | 92 | |
NM_001316557.1 | NP_001303486.1 | 2 | 584 | 92 | ||
NM_057869.4 | NP_477217.1 | 3 | 675 | 92 | ||
NM_169436.3 | NP_731641.1 | 1 | 646 | 92 | ||
GenBank mRNA | BT011037.1 | - | 3 | 674 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |