Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170138.1 | NP_001163609.1 | 6 - 7 | 868 | 54 | |
NM_001170142.1 | NP_001163613.1 | 6 - 7 | 802 | 54 | ||
NM_001275656.1 | NP_001262585.1 | 6 - 7 | 796 | 54 | ||
NM_169613.1 | NP_731970.1 | 6 - 7 | 802 | 54 | ||
GenBank mRNA | AY058374.1 | - | 6 - 7 | 857 | 54 | |
AY113622.1 | - | 6 - 7 | 803 | 54 | ||
BT081982.1 | - | 6 - 7 | 859 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |