Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cpo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014631.3 | NP_001014631.3 | 3 - 4 | 3060 | 58 | |
NM_001014632.4 | NP_001014632.3 | 3 - 4 | 1949 | 58 | ||
NM_001170169.3 | NP_001163640.2 | 3 - 4 | 1716 | 58 | ||
NM_001275759.1 | NP_001262688.1 | 3 - 4 | 1876 | 58 | ||
NM_001275760.1 | NP_001262689.1 | 3 - 4 | 1949 | 58 | ||
NM_001275761.1 | NP_001262690.1 | 3 - 4 | 2004 | 58 | ||
NM_001275762.1 | NP_001262691.1 | 3 - 4 | 2004 | 58 | ||
NM_001275763.1 | NP_001262692.1 | 3 - 4 | 1949 | 58 | ||
NM_080105.4 | NP_524844.6 | 3 - 4 | 1949 | 58 | ||
NM_169781.4 | NP_732282.6 | 3 - 4 | 3060 | 58 | ||
NM_169782.3 | NP_732283.5 | 3 - 4 | 2004 | 58 | ||
NM_169783.4 | NP_732284.5 | 3 - 4 | 1949 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |