Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ssh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170245.1 | NP_001163716.1 | 4 - 5 | 1017 | 106 | |
NM_001170246.1 | NP_001163717.1 | 4 - 5 | 1250 | 106 | ||
NM_079768.4 | NP_524492.2 | 4 - 5 | 1250 | 106 | ||
NM_170184.2 | NP_733063.1 | 4 - 5 | 1017 | 106 | ||
GenBank mRNA | AB036834.1 | - | 4 - 5 | 1250 | 106 | |
BT011408.1 | - | 4 - 5 | 1017 | 106 | ||
BT120385.1 | - | 4 - 5 | 1226 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |