Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276165.1 | NP_001263094.1 | 2 - 3 | 238 | 64 | |
NM_079833.2 | NP_524557.1 | 2 - 3 | 404 | 64 | ||
GenBank mRNA | AY119592.1 | - | 2 - 3 | 404 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |