Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp4c3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079859.4 | NP_524598.1 | 7 - 8 | 1394 | 62 | |
GenBank mRNA | BT010108.2 | - | 7 - 8 | 1471 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |