Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: boss | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080709.3 | NP_542440.1 | 3 - 4 | 701 | 83 | |
GenBank mRNA | BT088800.1 | - | 3 - 4 | 686 | 83 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Drosophila melanogaster |