Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144658.2 | NP_001138130.2 | 4 - 5 | 2015 | 64 | |
NM_170531.4 | NP_733410.2 | 4 - 5 | 2015 | 64 | ||
NM_206591.2 | NP_996314.1 | 4 - 5 | 2030 | 64 | ||
GenBank mRNA | BT009990.1 | - | 4 - 5 | 2013 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |