Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276121.1 | NP_001263050.1 | 5 - 6 | 1078 | 60 | |
NM_001300646.1 | NP_001287575.1 | 5 - 6 | 1125 | 60 | ||
NM_079991.3 | NP_524730.1 | 5 - 6 | 1078 | 60 | ||
GenBank mRNA | AF172258.1 | - | 5 - 6 | 1057 | 60 | |
AY058643.1 | - | 5 - 6 | 1051 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster Inventoried |