Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snRNP-U1-70K | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273244.1 | NP_001260173.1 | 2 | 1643 | 82 | |
NM_057857.5 | NP_477205.1 | 2 | 1643 | 82 | ||
GenBank mRNA | AY061501.1 | - | 2 | 1630 | 82 | |
AY089622.1 | - | 2 | 1642 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |