Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300350.1 | NP_001287279.1 | 2 | 1157 | 73 | |
NM_001300351.1 | NP_001287280.1 | 2 | 857 | 73 | ||
NM_079588.3 | NP_524312.1 | 2 | 1164 | 73 | ||
NM_169373.2 | NP_731529.1 | 2 | 850 | 73 | ||
GenBank mRNA | AB050622.1 | - | 1 | 608 | 73 | |
BT015244.1 | - | 2 | 1164 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |