Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RpS15Aa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169267.2 | NP_001162738.1 | 3 | 470 | 70 | |
NM_079970.6 | NP_524709.1 | 2 | 537 | 70 | ||
NM_167360.3 | NP_727690.1 | 3 | 474 | 70 | ||
NM_167362.3 | NP_727692.1 | 3 | 533 | 70 | ||
NM_167363.3 | NP_727693.1 | 2 | 537 | 70 | ||
GenBank mRNA | AY084182.1 | - | 3 | 482 | 70 | |
Z21673.1 | - | 459 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |