Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jon25Bi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038784.2 | NP_001033873.1 | 1 | 813 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |