Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hacd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199861.1 | NP_956155.1 | 2 - 3 | 296 | 74 | |
GenBank mRNA | BC044396.1 | - | 2 - 3 | 296 | 74 | |
BC165084.1 | - | 2 - 3 | 258 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |