Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pygmb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328031.1 | NP_001314960.1 | 4 - 5 | 686 | 85 | |
XM_017356828.2 | XP_017212317.1 | 4 - 5 | 527 | 85 | ||
GenBank mRNA | BC055181.1 | - | 4 - 5 | 686 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |