Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rer1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200675.2 | NP_956969.1 | 3 - 4 | 223 | 86 | |
XM_005167296.4 | XP_005167353.2 | 3 - 4 | 305 | 86 | ||
XM_005167298.4 | XP_005167355.2 | 3 - 4 | 290 | 86 | ||
XM_005167299.4 | XP_005167356.1 | 3 - 4 | 310 | 86 | ||
GenBank mRNA | AY398406.1 | - | 3 - 4 | 223 | 86 | |
BC050162.1 | - | 3 - 4 | 315 | 86 | ||
BC058292.1 | - | 3 - 4 | 225 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |