Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ubr7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212629.1 | NP_997794.1 | 1 - 2 | 325 | 94 | |
XM_021468365.1 | XP_021324040.1 | 1 - 2 | 737 | 94 | ||
GenBank mRNA | BC044390.1 | - | 1 - 2 | 325 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |