Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: quo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004109.2 | NP_001004109.2 | 22 - 23 | 6144 | 97 | |
GenBank mRNA | AY654285.2 | - | 22 - 23 | 6149 | 97 | |
BC162633.1 | - | 22 - 23 | 6070 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |