Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tnk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080017.2 | NP_001073486.2 | 10 - 11 | 1476 | 103 | |
XM_005172298.4 | XP_005172355.1 | 10 - 11 | 1694 | 103 | ||
XM_017356886.2 | XP_017212375.1 | 9 - 10 | 1596 | 103 | ||
GenBank mRNA | BC125935.1 | - | 10 - 11 | 1476 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |