Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: map2k4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001089421.1 | NP_001082890.1 | 9 - 10 | 867 | 99 | |
XM_005172951.4 | XP_005173008.1 | 8 - 9 | 1218 | 99 | ||
XM_021480155.1 | XP_021335830.1 | 9 - 10 | 1255 | 99 | ||
XM_021480156.1 | XP_021335831.1 | 8 - 9 | 1223 | 99 | ||
GenBank mRNA | AB438979.1 | - | 9 - 10 | 867 | 99 | |
BC162777.1 | - | 9 - 10 | 943 | 99 | ||
EF489398.1 | - | 9 - 10 | 867 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |