Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ednrba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131197.1 | NP_571272.1 | 3 - 4 | 880 | 71 | |
GenBank mRNA | AF275636.1 | - | 3 - 4 | 880 | 71 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Detect Genomic DNA Danio rerio Probe spans exons |