Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pcbd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200814.1 | NP_957108.1 | 3 - 4 | 238 | 98 | |
GenBank mRNA | AF473825.1 | - | 2 - 3 | 219 | 98 | |
BC059694.1 | - | 3 - 4 | 238 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |