Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dcdc2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037689.2 | NP_001032778.2 | 11 - 12 | 1177 | 79 | |
XM_017358759.2 | XP_017214248.1 | 13 - 14 | 1564 | 79 | ||
XM_017358760.2 | XP_017214249.1 | 13 - 14 | 1538 | 79 | ||
XM_017358761.2 | XP_017214250.1 | 13 - 14 | 1551 | 79 | ||
XM_017358762.2 | XP_017214251.1 | 13 - 14 | 1525 | 79 | ||
XM_017358763.2 | XP_017214252.1 | 13 - 14 | 1543 | 79 | ||
XM_017358764.2 | XP_017214253.1 | 13 - 14 | 1504 | 79 | ||
XM_017358765.2 | XP_017214254.1 | 13 - 14 | 1564 | 79 | ||
XM_021480609.1 | XP_021336284.1 | 12 - 13 | 2938 | 79 | ||
GenBank mRNA | BC108062.1 | - | 11 - 12 | 1188 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |