Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc93a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044916.1 | NP_001038381.1 | 6 - 7 | 965 | 67 | |
XM_005170182.3 | XP_005170239.1 | 6 - 7 | 1120 | 67 | ||
GenBank mRNA | BC171433.1 | - | 6 - 7 | 1021 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |