Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slmapa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077254.1 | NP_001070722.1 | 1 - 2 | 685 | 71 | |
XM_005168060.4 | XP_005168117.1 | 1 - 2 | 1378 | 71 | ||
XM_005168061.4 | XP_005168118.1 | 1 - 2 | 1378 | 71 | ||
XM_005168062.4 | XP_005168119.1 | 1 - 2 | 1378 | 71 | ||
XM_005168063.4 | XP_005168120.1 | 1 - 2 | 1378 | 71 | ||
GenBank mRNA | BC124763.1 | - | 1 - 2 | 685 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |