Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cthrc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003765.2 | NP_001003765.1 | 2 - 3 | 409 | 92 | |
GenBank mRNA | BC079494.1 | - | 2 - 3 | 413 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |