Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: elovl1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005989.3 | NP_001005989.1 | 5 - 6 | 694 | 82 | |
XM_005171196.4 | XP_005171253.1 | 5 - 6 | 738 | 82 | ||
GenBank mRNA | BC083417.1 | - | 5 - 6 | 450 | 82 | |
EH495486.1 | - | 5 - 6 | 636 | 82 | ||
EH549422.1 | - | 5 - 6 | 735 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |