Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bin2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007421.1 | NP_001007422.1 | 7 - 8 | 674 | 80 | |
XM_009296976.3 | XP_009295251.1 | 7 - 8 | 768 | 80 | ||
XM_009296977.3 | XP_009295252.1 | 7 - 8 | 821 | 80 | ||
GenBank mRNA | BC085363.1 | - | 7 - 8 | 674 | 80 | |
BC165739.1 | - | 6 - 7 | 561 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |