Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: herpud1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025537.1 | NP_001020708.1 | 1 - 2 | 281 | 79 | |
XM_005159120.4 | XP_005159177.1 | 1 - 2 | 366 | 79 | ||
XM_005159121.4 | XP_005159178.1 | 1 - 2 | 340 | 79 | ||
GenBank mRNA | BC097190.1 | - | 1 - 2 | 281 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |