Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zp3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131696.1 | NP_571771.1 | 4 - 5 | 925 | 92 | |
GenBank mRNA | BC067692.1 | - | 5 - 6 | 875 | 92 | |
BC151892.1 | - | 4 - 5 | 927 | 92 | ||
U55863.3 | - | 4 - 5 | 925 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |