Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
TaqMan
®
Gene Expression Assays
›
Search Tool
›
Search Results
› Dr03094781_g1
See other TCEA3 GE Assays ›
Gene Symbol:
tcea3
Gene Name:
transcription elongation factor A (SII), 3
Gene Aliases:
zgc:77732
Chromosome Location:
Chr.16: 34111892 - 34135268 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID
Dr03094781_g1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: tcea3
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_205683.1
NP_991246.1
6 - 7
624
92
XM_005158337.4
XP_005158394.1
18 - 19
1783
92
XM_009292441.3
XP_009290716.1
15 - 16
1615
92
XM_009292443.3
XP_009290718.1
14 - 15
1561
92
XM_009292444.3
XP_009290719.1
16 - 17
1521
92
XM_009292445.3
XP_009290720.1
13 - 14
1506
92
XM_009292446.3
XP_009290721.1
16 - 17
1485
92
XM_009292449.3
XP_009290724.1
9 - 10
1003
92
XM_009292450.3
XP_009290725.1
8 - 9
919
92
XM_009292451.3
XP_009290726.1
9 - 10
795
92
XM_009292452.3
XP_009290727.1
6 - 7
627
92
XM_009292453.3
XP_009290728.1
5 - 6
573
92
XM_017351328.2
XP_017206817.1
17 - 18
1729
92
XM_017351329.2
XP_017206818.1
17 - 18
1723
92
XM_021466657.1
XP_021322332.1
17 - 18
1729
92
XM_021466659.1
XP_021322334.1
17 - 18
1729
92
XM_021466660.1
XP_021322335.1
16 - 17
1675
92
XM_021466661.1
XP_021322336.1
16 - 17
1675
92
XM_021466662.1
XP_021322337.1
16 - 17
1669
92
XM_021466663.1
XP_021322338.1
16 - 17
1669
92
XM_021466664.1
XP_021322339.1
14 - 15
1560
92
XM_021466665.1
XP_021322340.1
16 - 17
1491
92
GenBank mRNA
BC065444.1
-
6 - 7
624
92
Back To Top
More Information
Set Membership:
Amplicon length between 86 and 100
Mammalian Gene Collection
Detect Genomic DNA
Danio rerio
Probe spans exons
Panther Classification:
Molecular Function -
general transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
translation
translational elongation
nucleic acid binding
translation elongation factor activity
zinc ion binding
metal ion binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-f6n66:80/10.239.161.116:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0