Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cab39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002145.1 | NP_001002145.1 | 8 - 9 | 1043 | 87 | |
GenBank mRNA | BC071393.1 | - | 8 - 9 | 1043 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |