Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sncgb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020652.1 | NP_001018488.1 | 1 - 2 | 264 | 84 | |
XM_021480112.1 | XP_021335787.1 | 1 - 2 | 84 | |||
GenBank mRNA | BC095672.1 | - | 1 - 2 | 264 | 84 | |
EU368946.1 | - | 1 - 2 | 336 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |